Läs mer
Tranylcypromine Hydrochloride, TRC
Produktkod. 30136998
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136998 25 mg
30008594 5 mg
30005957 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136998 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT71500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4548-34-9
Molekylformel C9 H12 Cl N
Molekylvikt (g/mol) 169.65
Synonym Cyclopropanamine, 2-phenyl-, hydrochloride (1:1), (1S,2R)- (ACI), Cyclopropanamine, 2-phenyl-, hydrochloride, (1S,2R)- (9CI), Cyclopropanamine, 2-phenyl-, hydrochloride, (1S-trans)- (ZCI), Cyclopropylamine, 2-phenyl-, hydrochloride, trans-(+)- (8CI), (1S,2R)-2-Phenylcyclopropanamine hydrochloride, 1-(S)-Amino-2-(R)-phenylcyclopropane hydrochloride, d-Tranylcypromine hydrochloride, Tranylcypromine hydrochloride, (+)-Tranylcypromine hydrochloride, (1S,2R)-Tranylcypromine hydrochloride
IUPAC-namn (1S,2R)-2-phenylcyclopropan-1-amine;hydrochloride
LEDER Cl.N[C@H]1C[C@@H]1c2ccccc2
Specifikationer
Kemiskt namn eller material Tranylcypromine Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C9H11N.ClH/c10-9-6-8(9)7-4-2-1-3-5-7;/h1-5,8-9H,6,10H2;1H/t8-,9+;/m1./s1
Formel vikt 169.0658