missing translation for 'onlineSavingsMsg'
Learn More

TRAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10325-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826643

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The 90 kDa heat shock protein (hsp90) family of molecular chaperones is a highly conserved family of proteins that play an important physiological role. Hsp90 is involved in numerous cellular processes but is best known for its association with signal transduction machinery. A recently cloned homolog of hsp90 is the tumor necrosis factor receptor-associated protein (TRAP1). Like hsp90, TRAP1 is found to be associated with numerous proteins involved in diverse actions. Immunofluorescence data has shown TRAP1 to be localized in the mitochondria of mammalian cells. This observation and the fact that TRAP1 is shown to have a mitochondrial targeting presequence strongly implicates TRAP1 as a mitochondrial matrix protein.
Specifications

Specifications

TRAP1
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12931, Q5XHZ0, Q9CQN1
HSP 75, HSP75, HSP90L, TNFR associated protein 1, TRAP 1, TRAP1
Rabbit
IgG
20 μL
-20°C
Primary
10131, 287069, 68015
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
TRAP1
Liquid
Trap1
TRAP1 Fusion Protein Ag0400
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers