missing translation for 'onlineSavingsMsg'
Learn More

TRAPB/SSR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10278-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826563

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23.
Specifications

Specifications

TRAPB/SSR2
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P43308, Q9CPW5
HSD25, SSR beta, SSR2, TLAP, TRAP BETA, TRAPB
Rabbit
IgG
150 μL
-20°C
Primary
295235, 66256, 6746
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SSR2
Liquid
Ssr2
TRAPB/SSR2 Fusion Protein Ag0304
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali