Läs mer
Traumatic Acid, TRC
Produktkod. 30452256
Change view
Kvantitet:
100 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452256 500 mg
30450737 10 g
30455902 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30452256 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT715500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6402-36-4
Molekylformel C12H20O4
Molekylvikt (g/mol) 228.28
Synonym (E)-2-Dodecenedioic Acid, trans-2-Dodecenedioic Acid, trans-Traumatic Acid
IUPAC-namn (E)-dodec-2-enedioic acid
LEDER OC(=O)CCCCCCCC\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Traumatic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C12H20O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h7,9H,1-6,8,10H2,(H,13,14)(H,15,16)/b9-7+
Formel vikt 228.1362