Läs mer
Trazodone N-Oxide, TRC
Produktkod. 17992265
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17992265 50 mg
17964315 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17992265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT71851050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55290-68-1
Molekylformel C19H22ClN5O2
Molekylvikt (g/mol) 387.86
Synonym 2-[3-[4-(3-Chlorophenyl)-1-oxido-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one;
LEDER ClC1=CC(N2CCN(CC2)(CCCN3N=C(C=CC=C4)N4C3=O)=O)=CC=C1
Specifikationer
Kemiskt namn eller material Trazodone N-Oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C19H22ClN5O2/c20-16-5-3-6-17(15-16)22-10-13-25(27,14-11-22)12-4-9-24-19(26)23-8-2-1-7-18(23)21-24/h1-3,5-8,15H,4,9-14H2
Formel vikt 387.1462
Procent renhet >95%