Läs mer
Trehalose 6-Phosphate Dipotassium Salt, TRC
Produktkod. 30456241
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30456241 2.5 mg
1 options
Produktkod. 30456241 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7187152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trehalose 6-Phosphate Dipotassium Salt
CAS 136632-28-5
Molekylformel C12 H21 K2 O14 P
Kvantitet 2.5 mg
Synonym α-D-Glucopyranosyl α-D-Glucopyranoside 6-(Dihydrogen Phosphate) Potassium Salt, T6P, Trehalose 6-Phosphate Potassium Salt
InChI formel InChI=1S/C12H23O14P.2K/c13-1-3-5(14)7(16)9(18)11(24-3)26-12-10(19)8(17)6(15)4(25-12)2-23-27(20,21)22;;/h3-19H,1-2H2,(H2,20,21,22);;/q;2*+1/p-2/t3-,4-,5-,6-,7+,8+,9?,10-,11+,12+;;/m1../s1
LEDER OC[C@H]1O[C@@H](O[C@@H]2O[C@H](COP(=O)(O[K])O[K])[C@@H](O)[C@H](O)[C@H]2O)C(O)[C@@H](O)[C@@H]1O
IUPAC-namn [potassiooxy-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(2S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]methoxy]phosphoryl]oxypotassium
Molekylvikt (g/mol) 498.46
Formel vikt 497.9943