Läs mer
Trenbolone cyclohexylmethylcarbonate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Trenbolone Cyclohexylmethylcarbonate |
| CAS | 23454-33-3 |
| Analyt- eller komponentnamn | Trenbolone Cyclohexylmethylcarbonate |
| Frakt skick | +20°C |
| Molekylformel | C26 H34 O4 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), Estra-4,9,11-trien-3-one, 17-[[(cyclohexylmethoxy)carbonyl]oxy]-, (17β)-, Estra-4,9,11-trien-3-one, 17β-hydroxy-, cyclohexylmethyl carbonate (8CI), Carbonic acid, cyclohexylmethyl ester, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Cyclohexanemethanol, hydrogen carbonate, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Parabolan, RU 2580, Trenbolone cyclohexylmethyl carbonate, Trenbolone hexahydrobenzyl carbonate |
| InChI formel | InChI=1S/C26H34O4/c1-26-14-13-21-20-10-8-19(27)15-18(20)7-9-22(21)23(26)11-12-24(26)30-25(28)29-16-17-5-3-2-4-6-17/h13-15,17,22-24H,2-12,16H2,1H3/t22-,23+,24+,26+/m1/s1 |
| LEDER | C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2OC(=O)OCC5CCCCC5 |
| Visa mer |