Learn More
Uvex Trend Black/Blue PU 69351 Boot - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  Uvex 6935150
Additional Details : Weight : 1.99000kg
 View more versions of this product
Product Code. 17203681
  • 1255.00 SEK / 1 pair
Estimated Shipment: 04-01-2024
to see stock.
Add to basket
Description
Description
Uvex 2 Trend Boot 69351 S3 SRC
  • Sporty S3 safety boot with wide fit
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Smooth-running, dirt and heat-resistant lacing system
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Penetration-resistant steel midsole, steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Foamed polyurethane scuffcap helps increase product life of upper
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding collar and dust tongue
  • Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69351
  • Standard EN ISO 20345:2011 S3 SRC
  • Sizes: 38 to 52
Specifications
Specifications
Black, Blue
50
Steel
EN ISO 20345:2011
Narrow width
Yes
S3
Leather
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC