Learn More
Uvex Trend Black/Blue PU 69357 Boot - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1225.00 SEK
Specifications
Color | Black, Blue |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17233711
|
Uvex
6935738 |
38 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17243711
|
Uvex
6935739 |
39 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17253711
|
Uvex
6935740 |
40 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17263711
|
Uvex
6935741 |
41 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17273711
|
Uvex
6935742 |
42 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17283711
|
Uvex
6935743 |
43 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17293711
|
Uvex
6935744 |
44 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17203721
|
Uvex
6935745 |
45 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17213721
|
Uvex
6935746 |
46 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17223721
|
Uvex
6935747 |
47 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17233721
|
Uvex
6935748 |
48 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17243721
|
Uvex
6935749 |
49 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17253721
|
Uvex
6935750 |
50 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17263721
|
Uvex
6935751 |
51 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17273721
|
Uvex
6935752 |
52 |
1225.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Boot 69357 S2 SRC
- Sporty S2 safety boot with wide fit
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Smooth-running, dirt and heat-resistant lacing system
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole
- Moisture transport system helps maintain foot temperature
- Soft padding collar and dust tongue
- Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69357
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Steel | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |