Learn More
Uvex Trend Black/Blue PU/PU 69348 Low Shoe - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  Uvex 6934843
706.67 SEK valid until 2023-05-26
Use promo code "20831" to get your promotional price.
Additional Details : Weight : 1.29500kg
 View more versions of this product
Product Code. 17233641
  • 1060.00 SEK / 1 pair
Estimated Shipment: 28-06-2023
to see stock.
Add to basket
Description
Description
Uvex 2 Trend Shoe 69348 S2 SRC
  • Sporty S2 safety shoe with wide fit
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Smooth-running, dirt and heat-resistant lacing system
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding collar and dust tongue
  • Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69348
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 38 to 52
Especificaciones
Especificaciones
Black, Blue
43
Steel
EN ISO 20345:2011
Standard width
Yes
S2
Leather
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC
Videos
SDS
Documentos
Documentos
Product Certifications
Ofertas especiales
Ofertas especiales