Learn More
Uvex Trend Black/Blue PU/PU 69397 Low Shoe - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1135.00 SEK
Specifications
Color | Black, Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17283931
|
Uvex
6939738 |
38 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17293931
|
Uvex
6939739 |
39 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17203941
|
Uvex
6939740 |
40 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17213941
|
Uvex
6939741 |
41 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17223941
|
Uvex
6939742 |
42 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17233941
|
Uvex
6939743 |
43 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17243941
|
Uvex
6939744 |
44 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17253941
|
Uvex
6939745 |
45 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17263941
|
Uvex
6939746 |
46 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17273941
|
Uvex
6939747 |
47 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17283941
|
Uvex
6939748 |
48 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17293941
|
Uvex
6939749 |
49 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17203951
|
Uvex
6939750 |
50 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17213951
|
Uvex
6939751 |
51 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17223951
|
Uvex
6939752 |
52 |
1135.00 SEK
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Shoe 69397 S2 SRC
- Sporty S2 safety shoe with wide fit
- Breathable, water repellent microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable insole
- Moisture transport system helps maintain foot temperature
- Soft padding on collar and dust tongue
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69397
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |