Learn More
Uvex Trend Black/Blue PU/PU 69399 Low Shoe - Width 12
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1135.00 SEK
Specifications
Color | Black, Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17283961
|
Uvex
6939938 |
38 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17293961
|
Uvex
6939939 |
39 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17203971
|
Uvex
6939940 |
40 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17213971
|
Uvex
6939941 |
41 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17223971
|
Uvex
6939942 |
42 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17233971
|
Uvex
6939943 |
43 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17243971
|
Uvex
6939944 |
44 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17253971
|
Uvex
6939945 |
45 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17263971
|
Uvex
6939946 |
46 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17273971
|
Uvex
6939947 |
47 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17283971
|
Uvex
6939948 |
48 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17293971
|
Uvex
6939949 |
49 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17203981
|
Uvex
6939950 |
50 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17213981
|
Uvex
6939951 |
51 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17223981
|
Uvex
6939952 |
52 |
1135.00 SEK
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Shoe 69399 S2 SRC
- Sporty S2 safety shoe with wide fit
- Breathable, water repellent microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable insole
- Moisture transport system helps maintain foot temperature
- Soft padding on collar and dust tongue
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69399
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S2 |