Learn More
Uvex Trend Black/Gray/Red PU/PU 69063 Sandal - Width 12
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  Uvex 6906343
Additional Details : Weight : 1.25200kg
 View more versions of this product
Product Code. 17203231
  • 1010.00 SEK / 1 pair
Estimated Shipment: 25-10-2023
to see stock.
Add to basket
Description
Description
Uvex 2 Trend Sandal 6906.3 S1 P SRC
  • Sporty, S1 P safety sandal with wide fit
  • Breathable microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Double, individually adjustable counter-traction hook and loop straps
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Penetration-resistent steel midsole, steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
    • Helps maintain foot temperature for optimum wearer comfort
  • Soft padding on collar and tongue
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 6906.3
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
Specifications
Specifications
Black, Gray, Red
43
Steel
EN ISO 20345:2011
Wide width
Yes
S1
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC