Learn More
Uvex Trend Black/Gray/Red PU/PU 69078 Low Shoe - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1105.00 SEK
Specifications
Color | Black, Gray, Red |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17253251
|
Uvex
6907838 |
38 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17263251
|
Uvex
6907839 |
39 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17273251
|
Uvex
6907840 |
40 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17283251
|
Uvex
6907841 |
41 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17293251
|
Uvex
6907842 |
42 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17203261
|
Uvex
6907843 |
43 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17213261
|
Uvex
6907844 |
44 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17223261
|
Uvex
6907845 |
45 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17233261
|
Uvex
6907846 |
46 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17243261
|
Uvex
6907847 |
47 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17253261
|
Uvex
6907848 |
48 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17263261
|
Uvex
6907849 |
49 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17273261
|
Uvex
6907850 |
50 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17283261
|
Uvex
6907851 |
51 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17293261
|
Uvex
6907852 |
52 |
1105.00 SEK
1 pair |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Perforated Shoe 6907.8 S1 SRC
- Sporty, S1 perforated safety shoe with wide fit
- Breathable perforated microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
- Helps maintain foot temperature for optimum wearer comfort
- Soft padding on collar and tongue
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Article No. 6907.8
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 38 to 52
Specifications
Black, Gray, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S1 |