Learn More
Uvex Trend Black/Gray/Red PU/PU 69079 Low Shoe - Width 12
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1105.00 SEK
Specifications
Color | Black, Gray, Red |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17203271
|
Uvex
6907938 |
38 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17213271
|
Uvex
6907939 |
39 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17223271
|
Uvex
6907940 |
40 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17233271
|
Uvex
6907941 |
41 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17243271
|
Uvex
6907942 |
42 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17253271
|
Uvex
6907943 |
43 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17263271
|
Uvex
6907944 |
44 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17273271
|
Uvex
6907945 |
45 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17283271
|
Uvex
6907946 |
46 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17293271
|
Uvex
6907947 |
47 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17203281
|
Uvex
6907948 |
48 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17213281
|
Uvex
6907949 |
49 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17223281
|
Uvex
6907950 |
50 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17233281
|
Uvex
6907951 |
51 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17243281
|
Uvex
6907952 |
52 |
1105.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Perforated Shoe 6907.9 S1 SRC
- Sporty, S1 perforated safety shoe with wide fit
- Breathable perforated microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
- Helps maintain foot temperature for optimum wearer comfort
- Soft padding on collar and tongue
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Article No. 6907.9
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 38 to 52
Specifications
Black, Gray, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |