Learn More
Uvex Trend BOA™ Black/Blue PU/PU 69383 Low Shoe - Width 12
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1325.00 SEK
Specifications
Color | Black, Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17233921
|
Uvex
6938338 |
38 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17243921
|
Uvex
6938339 |
39 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17253921
|
Uvex
6938340 |
40 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17263921
|
Uvex
6938341 |
41 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17273921
|
Uvex
6938342 |
42 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17283921
|
Uvex
6938343 |
43 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17293921
|
Uvex
6938344 |
44 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17203931
|
Uvex
6938345 |
45 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17213931
|
Uvex
6938346 |
46 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17223931
|
Uvex
6938347 |
47 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17233931
|
Uvex
6938348 |
48 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17243931
|
Uvex
6938349 |
49 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17253931
|
Uvex
6938350 |
50 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17263931
|
Uvex
6938351 |
51 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17273931
|
Uvex
6938352 |
52 |
1325.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 69383 S1 P SRC
- Sporty S1 P perforated safety shoe with wide fit
- Breathable, perforated microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- BOA™ Fit System Dial for custom comfort that's consistent all day
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel midsole, steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable insole
- Moisture transport system helps maintain foot temperature
- Soft padding on collar and tongue
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69383
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |