Läs mer
Novus Biologicals™ TRERF1 Antibody [Allophycocyanin], Novus Biologicals™
Handla allt Bio Techne ProdukterBeskrivning
TRERF1 Polyclonal antibody specifically detects TRERF1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)
Specifikationer
Specifikationer
| Antigen | TRERF1 |
| Användningsområden | Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative) |
| Klassificering | Polyclonal |
| Konjugera | APC |
| Formulering | PBS |
| Gene Alias | BCAR2, Breast cancer estrogen resistance 2RAPAHSA277276, dJ139D8.5, FLJ21198, rapa-1, rapa-2, RP1-139D8.5, transcriptional regulating factor 1, transcriptional regulating protein 132, transcriptional-regulating factor 1, Transcriptional-regulating protein 132, TReP-132, TREP132, Zinc finger protein rapa, Zinc finger transcription factor TReP-132 |
| Värd art | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human transcriptional regulating protein 132 (transcriptional regulating factor 1) using the numbering given in entry NP_277037.1 (GeneID 55809). |
| Reningsmetod | Immunogen affinity purified |
| Kvantitet | 0.1 mL |
| Visa mer |