missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ tri-GalNAc biotin

Beskrivning
Tri-GalNAc-biotin is a biotinylated asialoglycoprotein receptor (ASGPR) ligand for lysosomal targeting chimera (LYTAC) research and development, comprising a triantennary N-acetylgalactosamine (GalNAc) joined by a PEG linker to biotin. Tri-GalNAc-biotin can induce ASGPR-mediated endocytosis and trafficking of fluorescently labeled neutravidin for lysosomal degradation in HepG2 cells in a time-dependent manner. The increase in fluorescent signal within cells plateaus after 16-20h, and this effect can be inhibited by addition of tri-GalNAc COOH (Cat. No. 7401).
Specifikationer
Specifikationer
| Kvantitet | 50 μg |
| Mål | Degrader Building Blocks |
| Molekylformel | C83H146N14O34S |
| Renhet | 0.9 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?