Läs mer
Tri-o-tolyl-Phosphine Oxide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tri-o-tolyl-Phosphine Oxide |
| CAS | 6163-63-9 |
| Molekylformel | C21 H21 O P |
| Kvantitet | 250 mg |
| Synonym | Tris(2-methylphenyl)phosphine Oxide, NSC 299861, Tri(o-methylphenyl)phosphine Monoxide, Tri-o-tolylphosphine Oxide, Eletriptan Impurity 14, Tris(2-tolyl)phosphine Oxide, Tris(o-tolyl)phosphine Oxide |
| InChI formel | InChI=1S/C21H21OP/c1-16-10-4-7-13-19(16)23(22,20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3/h4-15H,1-3H3 |
| LEDER | Cc1ccccc1P(=O)(c2ccccc2C)c3ccccc3C |
| IUPAC-namn | 1-bis(2-methylphenyl)phosphoryl-2-methylbenzene |
| Molekylvikt (g/mol) | 320.365 |
| Formel vikt | 320.133 |