Läs mer
Tri(Propylene Glycol Butyl Ether (Mixture of isomers), TRC
Produktkod. 30417739
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30417739 1 mL
1 options
Produktkod. 30417739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1849351ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[1-(1-butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol
CAS 55934-93-5
Molekylformel C13 H28 O4
Kvantitet 1 mL
Synonym Tri(Propylene Glycol Butyl Ether (Mixture of isomers), [2-(2-Butoxymethylethoxy)methylethoxy]propanol, 2-(2-Butoxymethylethoxy)methylethoxy)propanol, BFTG, Dalpad C, Dowanol TPnB, TPnB, Tripropylene Glycol Monobutyl Ether, Tripropylene Glycol n-Butyl Ether, Tripropyleneglycol Butyl Ether
InChI formel InChI=1S/C13H28O4/c1-5-6-7-15-9-12(3)17-10-13(4)16-8-11(2)14/h11-14H,5-10H2,1-4H3
LEDER CCCCOCC(C)OCC(C)OCC(C)O
IUPAC-namn 1-[1-(1-butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol
Molekylvikt (g/mol) 248.359
Formel vikt 248.199