Learn More
tri-Sodium Citrate Dihydrate, For analysis EMSURE™ ACS, MilliporeSigma™
Product Code. 15864820
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
Product Code. Quantity Packaging Unit Size
15864820 1 kg Plastic Bottle 1kg
1 options
Product Code. 15864820 Supplier Supelco Supplier No. 1064481000
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
I lager
Add to basket
Edge
Specifications
CAS 4-3-6132
Melting Point 150°C (Anhydrous substance)
Color White
pH 7.5 to 9.0 (50g/L, H2O, 25°C)
Odor Odorless
Packaging Plastic Bottle
Molecular Formula C6H9Na3O9
Quantity 1 kg
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Solubility Information 720g/L
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 294.10
PubChem CID 71474
ChEBI CHEBI:32142
Formula Weight 294.10
Percent Purity 99.0 to 101.0%
Grade ACS
Physical Form Powder
Show More Show Less
Product Identifier
  • Trisodium citrate dihydrate for analysis EMSURE® ACS,ISO,Reag. Ph Eur