missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
di-Sodium Hydrogen Phosphate Heptahydrate, ACS, EMSURE™, MilliporeSigma™
Specifications
Specifications
| Kemiskt namn eller material | di-Sodium Hydrogen Phosphate Heptahydrate |
| Smältpunkt | 48°C |
| CAS | 7782-85-6 |
| pH | 8.7 - 9.3 |
| Fysisk form | Solid |
| Kvantitet | 1 kg |
| Anteckningar om analysprocent | 98.0 - 102.0% |
| Molekylformel | H15Na2O11P |
| MDL-nummer | MFCD00149180 |
| Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?