Läs mer
Triacetyl-Beta-cyclodextrin, TRC
Produktkod. 30450209
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30450209 100 mg
1 options
Produktkod. 30450209 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT737000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triacetyl-beta-cyclodextrin
CAS 23739-88-0
Molekylformel C84H112O56
Kvantitet 100 mg
Synonym β-Cyclodextrin 2A,2B,2C,2D,2E,2F,2G,3A,3B,3C,3D,3E,3F,3G,6A,6B,6C,6D,6E,6F,6G-Heneicosaacetate, β-Cyclodextrin Heneicosaacetate, 2,4,7,9,12,14,17,19,22,24,27,29,32,34-Tetradecaoxaoctacyclo[31.2.2.23,6.28,11.213,16.218,21.223,26.228,31]nonatetracontane β-Cyclodextrin Deriv., 2,3,6-Triacetyl-β-cyclodextrin, Beta W 7 TA 3.0, Cavasol W7TA, Heptakis(2,3,6-tri-O-acetyl)-β-cyclodextrin, Peracetylated β-Cyclodextrin, β-Cyclodextrin Peracetate
InChI formel InChI=1S/C84H112O56/c1-29(85)106-22-50-57-64(113-36(8)92)71(120-43(15)99)78(127-50)135-58-51(23-107-30(2)86)129-80(73(122-45(17)101)65(58)114-37(9)93)137-60-53(25-109-32(4)88)131-82(75(124-47(19)103)67(60)116-39(11)95)139-62-55(27-111-34(6)90)133-84(77(126-49(21)105)69(62)118-41(13)97)140-63-56(28-112-35(7)91)132-83(76(125-48(20)104)70(63)119-42(14)98)138-61-54(26-110-33(5)89)130-81(74(123-46(18)102)68(61)117-40(12)96)136-59-52(24-108-31(3)87)128-79(134-57)72(121-44(16)100)66(59)115-38(10)94/h50-84H,22-28H2,1-21H3/t50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64?,65?,66?,67?,68?,69?,70?,71?,72?,73?,74?,75?,76?,77?,78-,79-,80-,81-,82-,83-,84+/m1/s1
LEDER O=C(C)O[C@H]([C@@H](OC(C)=O)[C@@H](O[C@H]1[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H](O[C@H]2[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H](O[C@H]3C(OC(C)=O)[C@@H](OC(C)=O)[C@H]4O[C@@H]3COC(C)=O)O[C@@H]2COC(C)=O)O[C@@H]1COC(C)=O)O[C@@H]5COC(C)=O)[C@@H]5O[C@@H](O[C@@H]6COC(C)=O)[C@H](OC(C)=O)C(OC(C)=O)[C@@H]6O[C@@H](O[C@@H]7COC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]7O[C@H]8O[C@H](COC(C)=O)[C@@H](O4)[C@H](OC(C)=O)[C@H]8OC(C)=O
Molekylvikt (g/mol) 2017.75
Formel vikt 2016.59