Läs mer
Triacontanoic Acid, TRC
Produktkod. 30454936
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454936 10 mg
30450047 2 mg
30453681 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454936 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89599810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 506-50-3
Molekylformel C30 H60 O2
Molekylvikt (g/mol) 452.8
Synonym Triacontanoic acid (6CI, 7CI, 8CI, 9CI, ACI), Melissic acid, Myricic acid, n-Triacontanoic acid, NSC 53832
IUPAC-namn triacontanoic acid
LEDER CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material Melissic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C30H60O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30(31)32/h2-29H2,1H3,(H,31,32)
Formel vikt 452.4593