Läs mer
Triadimefon, TRC
Produktkod. 30138317
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138317 5 g
30132941 25 g
17975025 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7671205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43121-43-3
Molekylformel C14 H16 Cl N3 O2
Molekylvikt (g/mol) 293.75
Synonym 2-Butanone, 1-(4-chlorophenoxy)-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)-, 1-(4-Chlorophenoxy)-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)-2-butanone, 1-(4-Chlorophenoxy)-3,3-dimethyl-1,2,4-triazol-1-yl-2-butanone, 1-(4-Chlorophenoxy)-3,3-dimethyl-1-(1,2,4-triazol-1-yl)-2-butanone, 1-(4-Chlorophenoxy)-3,3-dimethyl-1-(1-H-1,2,4-triazol-1-yl)butanone, Azocene, BAY 6681F, BAY-MEB 6447, Bayleton, Bayleton 25, Bayleton 250EC, Bayleton 5, Bayleton 5DF, Bayleton 5WP, Bayleton PB, Bayleton special, Fenxiunin, MEB 6447, NSC 303303, Tidifon, Triadimefon, Triadimefone, Triadimephon, Tripinacloraz, Webeton
IUPAC-namn 1-(4-chlorophenoxy)-3,3-dimethyl-1-(1,2,4-triazol-1-yl)butan-2-one
LEDER CC(C)(C)C(=O)C(Oc1ccc(Cl)cc1)n2cncn2
Specifikationer
Kemiskt namn eller material Triadimefon
Kvantitet 5 g
InChI formel InChI=1S/C14H16ClN3O2/c1-14(2,3)12(19)13(18-9-16-8-17-18)20-11-6-4-10(15)5-7-11/h4-9,13H,1-3H3
Formel vikt 293.0931