Läs mer
Triallate, TRC
Produktkod. 30450164
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30450164 250 mg
1 options
Produktkod. 30450164 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT767150250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triallate
CAS 2303-17-5
Anteckningar om renhetsgrad GC
Molekylformel C10 H16 Cl3 N O S
Kvantitet 250 mg
Synonym 2-Propene-1-thiol','2,3,3-trichloro-','diisopropylcarbamate (8CI), Carbamic acid','diisopropylthio-','S-(2,3,3-trichloroallyl) ester (7CI','8CI), Carbamothioic acid','bis(1-methylethyl)-','S-(2,3,3-trichloro-2-propenyl) ester (9CI), 2,3,3-Trichloroallyl N,N-diisopropylthiocarbamate, Avadex BW, CP 23426, Dipthal, Far-Go, MeSH ID: D014214, NSC 379698, S-2,3,3-Trichloroallyl diisopropylthiocarbamate, S-2,3,3-Trichloroallyl N,N-diisopropylthiocarbamate, Triallate, Triamyl, Trillate
InChI formel InChI=1S/C10H16Cl3NOS/c1-6(2)14(7(3)4)10(15)16-5-8(11)9(12)13/h6-7H,5H2,1-4H3
LEDER CC(C)N(C(C)C)C(=O)SCC(=C(Cl)Cl)Cl
IUPAC-namn S-(2,3,3-trichloroprop-2-enyl) N,N-di(propan-2-yl)carbamothioate
Molekylvikt (g/mol) 304.66
Formel vikt 303.0018