Läs mer
Triamcinolone Hexacetonide, TRC
Produktkod. 30134188
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134188 250 mg
30134149 1 g
17964685 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT767190250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5611-51-8
Molekylformel C30 H41 F O7
Molekylvikt (g/mol) 532.64
Synonym Triamcinolone Hexacetonide, Pregna-1,4-diene-3,20-dione, 21-(3,3-dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-(3,3-dimethylbutyrate) (8CI), (11β,16α)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Butyric acid, 3,3-dimethyl-, 21-ester with 9-fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione cyclic 16,17-acetal with acetone (8CI), Aristospan, CL 34433, Hexatrione, Lederlon, Lederspan, TATBA, Triamcinolone acetonide 21-tert-butylacetate, Triamcinolone acetonide tert-butyl acetate, Triamcinolone hexacetonide
IUPAC-namn (11β,16α)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
LEDER CC(C)(C)CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
Specifikationer
Kemiskt namn eller material Triamcinolone Hexacetonide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C30H41FO7/c1-25(2,3)15-24(35)36-16-22(34)30-23(37-26(4,5)38-30)13-20-19-9-8-17-12-18(32)10-11-27(17,6)29(19,31)21(33)14-28(20,30)7/h10-12,19-21,23,33H,8-9,13-16H2,1-7H3/t19-,20-,21-,23+,27-,28-,29-,30+/m0/s1
Formel vikt 532.2836
Procent renhet >95%