Läs mer
Triasulfuron, TRC
Produktkod. 30133664
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133664 250 mg
17957429 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT007645250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82097-50-5
Molekylformel C14 H16 Cl N5 O5 S
Molekylvikt (g/mol) 401.83
Synonym Benzenesulfonamide, 2-(2-chloroethoxy)-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]- (9CI, ACI), 2-(2-Chloroethoxy)-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide (ACI), Amber, Amber (insecticide), CGA 131036, Logran, Triasulfuron
IUPAC-namn 1-[2-(2-chloroethoxy)phenyl]sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
LEDER COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2OCCCl)n1
Specifikationer
Kemiskt namn eller material Triasulfuron
Kvantitet 250 mg
InChI formel InChI=1S/C14H16ClN5O5S/c1-9-16-12(19-14(17-9)24-2)18-13(21)20-26(22,23)11-6-4-3-5-10(11)25-8-7-15/h3-6H,7-8H2,1-2H3,(H2,16,17,18,19,20,21)
Formel vikt 401.0561