missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Mouse anti-Human, Mouse, Rat, Clone: 2C10C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66702-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.Specifications
TRIB3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K4K2, Q96RU7, Q9WTQ6 | |
TRIB3 | |
TRIB3 Fusion Protein Ag25776 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
2C10C7 | |
Unconjugated | |
TRIB3 | |
C20orf97, NIPK, SINK, SKIP3, TRB 3, TRB3, TRIB3, Tribbles homolog 3 | |
Mouse | |
Protein G | |
RUO | |
228775, 246273, 57761 | |
-20°C | |
Liquid |