Läs mer
Tribromoneopentanol, TRC
Produktkod. 30451077
Change view
Kvantitet:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451077 50 g
30454297 100 g
30451770 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30451077 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77264050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1522-92-5
Molekylformel C5 H9 Br3 O
Molekylvikt (g/mol) 324.84
Synonym 3-Bromo-2,2-bis(bromomethyl)-1-propanol, 2,2,2-Tris(bromomethyl)ethanol, 2,2-Bis(bromomethyl)-3-bromo-1-propanol, 3-Bromo-2,2-bis(bromomethyl)propanol, 3-Bromo-2,2-bis(bromomethyl)propyl alcohol, FR 1360, FR 513, NSC 20521, Pentaerythritol tribromide, Pentaerythritol tribromohydrin, Tribromoneopentanol, Tribromoneopentyl alcohol, 1-Propanol, 3-bromo-2,2-bis(bromomethyl)-
IUPAC-namn 3-bromo-2,2-bis(bromomethyl)propan-1-ol
LEDER OCC(CBr)(CBr)CBr
Specifikationer
Kemiskt namn eller material 2,2,2-Tris(bromomethyl)ethanol
Kvantitet 50 g
InChI formel InChI=1S/C5H9Br3O/c6-1-5(2-7,3-8)4-9/h9H,1-4H2
Formel vikt 321.8204