Läs mer
Tributyl (E)-Propene-1,2,3-tricarboxylate (Tributyl (E)-Aconitate), Mikromol™
Produktkod. 17929784
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
17929784 100 mg
1 options
Produktkod. 17929784 Leverantör Mikromol Leverantörsnummer MM0499.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tributyl (E)-Propene-1,2,3-tricarboxylate (Tributyl (E)-Aconitate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tributyl (E)-Propene-1,2,3-tricarboxylate (Tributyl (E)-Aconitate)
CAS 343599-72-4
Analyt- eller komponentnamn Tributyl (E)-Propene-1,2,3-tricarboxylate (Tributyl (E)-Aconitate)
Frakt skick Room Temperature
Molekylformel C18 H30 O6
Certifikat och efterlevnad ISO 9001
Synonym Tributyl Acetylcitrate Imp. B (EP) (E-Isomer), Tributyl (E)-Propene-1,2,3-tricarboxylate, Tributyl (E)-Aconitate, Tributyl Acetylcitrate Impurity B as (E)-Isomer
InChI formel InChI=1S/C18H30O6/c1-4-7-10-22-16(19)13-15(18(21)24-12-9-6-3)14-17(20)23-11-8-5-2/h13H,4-12,14H2,1-3H3/b15-13+
LEDER O=C(/C=C(C(OCCCC)=O)\CC(OCCCC)=O)OCCCC
IUPAC-namn tributyl (E)-prop-1-ene-1,2,3-tricarboxylate
Molekylvikt (g/mol) 342.43
Kvantitet 100 mg
Formel vikt 342.2042 g/mol