Läs mer
Tributyl(vinyl)tin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tributyl(vinyl)tin |
| CAS | 7486-35-3 |
| Molekylformel | C14 H30 Sn |
| Kvantitet | 10 g |
| Synonym | Stannane','tributylethenyl- (9CI','ACI), Stannane','tributylvinyl- (8CI), Tin','tributylvinyl- (6CI), Tributylethenylstannane (ACI), (Ethenyl)tributyltin, (Tributylstannyl)ethene, (Tributylstannyl)ethylene, Ethenyltributylstannane, Tributyl(vinyl)stannane, Tributylvinylstannane, Tributylvinyltin, Vinyltributylstannane, Vinyltributyltin |
| InChI formel | InChI=1S/3C4H9.C2H3.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1H,2H2; |
| LEDER | CCCC[Sn](CCCC)(CCCC)C=C |
| IUPAC-namn | tributyl(ethenyl)stannane |
| Molekylvikt (g/mol) | 317.1 |
| Formel vikt | 318.1369 |