Läs mer
Tributyltin Acrylate, TRC
Produktkod. 30450452
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450452 100 mg
30448937 1 g
30455270 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450452 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT774050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13331-52-7
Molekylformel C15H30O2Sn
Molekylvikt (g/mol) 361.1
Synonym Tributylacryloyloxystannane, Tributylstannyl Acrylate
IUPAC-namn tributylstannyl prop-2-enoate
LEDER CCCC[Sn](CCCC)(CCCC)OC(=O)C=C
Specifikationer
Kemiskt namn eller material Tributyltin Acrylate
Kvantitet 100 mg
InChI formel InChI=1S/3C4H9.C3H4O2.Sn/c3*1-3-4-2;1-2-3(4)5;/h3*1,3-4H2,2H3;2H,1H2,(H,4,5);/q;;;;+1/p-1
Formel vikt 362.1268