Learn More
Tributyltin Hydride (Stabilized), Sigma-Aldrich™
Product Code. 15834291
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834291 100 mL 100mL
15824291 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834291 Supplier Sigma Aldrich Supplier No. 8141090100
 Restricted Item
Beräknad leverans: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 688-73-3
Specifications
Density 1.10 g/cm3 at 20°C
Boiling Point 112°C to 114°C/11 hPa
MDL Number MFCD00009416
Quantity 100 mL
Synonym Tributyltin hydride (stabilized), Tributylstannane, Tributyltin hydride (stabilized)
Molecular Weight (g/mol) 291.06
Grade Research