missing translation for 'onlineSavingsMsg'
Läs mer

Tricaprin, TRC

Produktkod. 30454506
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30454506 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7740105G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Tricaprin
CAS 621-71-6
Molekylformel C33 H62 O6
Kvantitet 5 g
Synonym Decanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caprin, Decanoic acid 1,2,3-propanetriyl ester, tri-Decanoin, Actor M 2, Capric acid triglyceride, Capric triglyceride, Captex 1000, Decanoic acid triglyceride, Dynasan 110, Glycerin tricaprate, Glycerin tridecanoate, Glycerol tricaprate, Glycerol tricaprin, Glycerol tridecanoate, Glyceryl tricaprate, NSC 147475, Panacet 1000, Tri(decanoyl)glycerol, Tricapric glyceride, Tricaprin, Tridecanoin, Triglyceride DDD
InChI formel InChI=1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3
LEDER CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC
IUPAC-namn 2,3-di(decanoyloxy)propyl decanoate
Molekylvikt (g/mol) 554.84
Formel vikt 554.4546
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.