Läs mer
Tricaproin (>90%), TRC
Produktkod. 30449980
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30449980 10 mg
1 options
Produktkod. 30449980 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77378010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tricaproin
CAS 621-70-5
Molekylformel C21 H38 O6
Kvantitet 10 mg
Synonym Hexanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caproin, Hexanoic acid 1,2,3-propanetriyl ester, tri-Hexanoin, Caproic triglyceride, Glycerin tricaproate, Glycerin trihexanoate, Glycerol tricaproate, Glycerol tricapronate, Glyceryl tricaproate, NSC 406885, Tricaproin, Tricapronin, Tricaproylglycerol, Trihexanoin, Trihexanoylglycerol
InChI formel InChI=1S/C21H38O6/c1-4-7-10-13-19(22)25-16-18(27-21(24)15-12-9-6-3)17-26-20(23)14-11-8-5-2/h18H,4-17H2,1-3H3
LEDER CCCCCC(=O)OCC(COC(=O)CCCCC)OC(=O)CCCCC
IUPAC-namn 2,3-di(hexanoyloxy)propyl hexanoate
Molekylvikt (g/mol) 386.52
Formel vikt 386.2668