Läs mer
Trichlormethiazide, MedChemExpress
Produktkod. 16470838
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16470838 5 g 5g
16422109 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16470838 Leverantör Medchem Express Leverantörsnummer HYB0235
MedChemExpress Trichlormethiazide is an orally active thiazide diuretic, with antihypertensive effect. Trichlormethiazide increases urine volume (UV), Na and K excretion and tends to improve the depressed creatinine clearance (CCRE) in acute renal failure rats model.
Trichlormethiazide is an orally active thiazide diuretic, with antihypertensive effect. Trichlormethiazide increases urine volume (UV), Na and K excretion and tends to improve the depressed creatinine clearance (CCRE) in acute renal failure rats model.
Specifikationer
Kemiskt namn eller material Trichlormethiazide
CAS 133-67-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C8H8Cl3N3O4S2
Kvantitet 5 g
Löslighetsinformation DMSO : 150 mg/mL (394.05 mM; Need ultrasonic)
LEDER O=S(C1=C(Cl)C=C(C2=C1)NC(C(Cl)Cl)NS2(=O)=O)(N)=O
Molekylvikt (g/mol) 380.66
Formel vikt 380.66
Procent renhet 98.54%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-protein/nucleotide metabolism