Learn More
Trichloroacetonitrile, Sigma-Aldrich™
Product Code. 15894231
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894231 250 mL 250mL
15884231 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894231 Supplier Sigma Aldrich Supplier No. 8082220250
 Restricted Item
Beräknad leverans: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 545-06-2
Specifications
Melting Point -44°C
Density 1.44 g/cm3 at 20°C
Boiling Point 85.7°C/1013 hPa
MDL Number MFCD00001842
Quantity 250 mL
Synonym Trichloroacetonitrile, Trichloromethylnitrile, Trichloroacetonitrile
Molecular Weight (g/mol) 144.39
Grade Research