Läs mer
Trichloroethylene-d, TRC
Produktkod. 30454384
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30454384 50 mg
1 options
Produktkod. 30454384 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77403150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trichloroethylene-d
CAS 13291-68-4
Molekylformel C2 D Cl3
Kvantitet 50 mg
Synonym Ethene-d','trichloro- (9CI), Ethylene-d','trichloro- (6CI','7CI','8CI), 1,1,2-Trichloro-2-deuterioethene, Deuterotrichloroethylene
InChI formel InChI=1S/C2HCl3/c3-1-2(4)5/h1H/i1D
LEDER [2H]C(=C(Cl)Cl)Cl
IUPAC-namn 1,1,2-trichloro-2-deuterioethene
Molekylvikt (g/mol) 132.3945
Formel vikt 130.9207