Läs mer
Tricine, TRC
Produktkod. 30455351
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30455351 10 g
1 options
Produktkod. 30455351 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77365310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tricine
CAS 5704-04-1
Molekylformel C6H13NO5
Kvantitet 10 g
Synonym N-[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]glycine, N-[Tris(hydroxymethyl)methyl]glycine, NSC 369995
InChI formel InChI=1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)
LEDER OCC(CO)(CO)NCC(=O)O
IUPAC-namn 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
Molekylvikt (g/mol) 179.17
Formel vikt 179.0794