Läs mer
Triclosan Methyl Ether, TRC
Produktkod. 30451628
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30451628 50 mg
1 options
Produktkod. 30451628 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77427550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triclosan methyl ether
CAS 4640-01-1
Molekylformel C13 H9 Cl3 O2
Kvantitet 50 mg
Synonym Benzene','4-chloro-1-(2,4-dichlorophenoxy)-2-methoxy-, 4-Chloro-1-(2,4-dichlorophenoxy)-2-methoxybenzene, 2,4,4'-Trichloro-2'-methoxydiphenyl ether, 5-Chloro-2-(2,4-dichlorophenoxy)anisole, Methyl triclosan, Triclosan methyl, Triclosan-Me, Triclosan methyl ether, Triclosan-methyl ether
InChI formel InChI=1S/C13H9Cl3O2/c1-17-13-7-9(15)3-5-12(13)18-11-4-2-8(14)6-10(11)16/h2-7H,1H3
LEDER COc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl
IUPAC-namn 2,4-dichloro-1-(4-chloro-2-methoxyphenoxy)benzene
Molekylvikt (g/mol) 303.57
Formel vikt 301.9668