Learn More
Cytiva Tricorn™ Filter Kits
Filter kits for Tricorn™ 5 and 10 columns
1199.00 SEK - 1379.00 SEK
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
15201228
|
Cytiva
29053586 |
For 5 columns |
1379.00 SEK
1 set |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10660561
|
Cytiva
11-0012-54 |
For 10 columns |
1199.00 SEK
1 set |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
Kit includes:
- Top and bottom adapters
- Bottom filters
- EPDM O-rings