Läs mer
Tricyclene, TRC
Produktkod. 30455078
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455078 50 mg
30455079 100 mg
30450063 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79746550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 508-32-7
Molekylformel C10H16
Molekylvikt (g/mol) 136.24
Synonym 1,7,7-Trimethyltricyclo[2.2.1.02,6]heptane, Cyclene, NSC 86978, Teresantanane
IUPAC-namn 6,7,7-trimethyl-2,3,4,5-tetrahydro-1H-tricyclo[2.2.1.0^{2,6}]heptane
LEDER CC1(C)C2CC3C(C2)C13C
Specifikationer
Kemiskt namn eller material Tricyclene
Kvantitet 50 mg
InChI formel InChI=1S/C10H16/c1-9(2)6-4-7-8(5-6)10(7,9)3/h6-8H,4-5H2,1-3H3
Formel vikt 136.1252