Läs mer
Tricyclopentadiene, TRC
Produktkod. 30179755
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179755 100 mg
30180330 500 mg
30180195 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT778338100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7158-25-0
Molekylformel C15H18
Molekylvikt (g/mol) 198.3
Synonym 3a,4,4a,5,8,8a,9,9a-Octahydro-4,9:5,8-dimethano-1H-benz[f]indene;3a,4,4a,5,8,8a,9,9a-Octahydro-4,9:5,8-dimethano-1H-cyclopenta[a]naphthalene, 4,9:5,8-Dimethano-3a,4,4a,5,8,8a,9,9a-octahydro-1H-benzindene, Cyclopentadiene trimer, DMOB, Pentacyclo[6.5.1.02,7.13,6.09,13]pentadeca-4,10-diene
LEDER O[C@H](CCCC1)[C@@H]1C2=CC=CC=C2
Specifikationer
Kemiskt namn eller material Tricyclopentadiene
Kvantitet 100 mg
InChI formel InChI=1S/C15H18/c1-2-10-11(3-1)13-7-12(10)14-8-4-5-9(6-8)15(13)14/h1-2,4-5,8-15H,3,6-7H2