Läs mer
Tridecafluorohexanesulfonic Acid Potassium Salt (Mixture of Isomers), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Perfluorohexanesulfonic acid potassium salt |
| CAS | 3871-99-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C6 F13 O3 S . K |
| Kvantitet | 250 mg |
| Synonym | Perfluorohexane Sulfonic Acid Potassium, Potassium perfluorohexanesulfonate, 1-Hexanesulfonic acid','1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-','potassium salt (9CI), 1-Hexanesulfonic acid','tridecafluoro-','potassium salt (6CI','8CI), Potassium (perfluorohexyl)sulfonate, Potassium perfluoro-1-hexanesulfonate, Potassium perfluorohexanesulfonate, Potassium tridecafluorohexan-1-sulfonate, Potassium tridecafluorohexane-1-sulfonate, Tridecafluorohexane-1-sulfonic acid potassium salt, Tridecafluorohexanesulfonic acid potassium salt |
| InChI formel | InChI=1S/C6HF13O3S.K/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22;/h(H,20,21,22);/q;+1/p-1 |
| LEDER | [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | potassium;1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonate |
| Molekylvikt (g/mol) | 438.2 |
| Visa mer |