missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tridecafluorohexanesulfonic Acid Potassium Salt (Mixture of Isomers), TRC
Specifications
Specifications
| Kemiskt namn eller material | Perfluorohexanesulfonic acid potassium salt |
| CAS | 3871-99-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C6 F13 O3 S . K |
| Kvantitet | 50 mg |
| Synonym | Perfluorohexane Sulfonic Acid Potassium, Potassium perfluorohexanesulfonate, 1-Hexanesulfonic acid','1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-','potassium salt (9CI), 1-Hexanesulfonic acid','tridecafluoro-','potassium salt (6CI','8CI), Potassium (perfluorohexyl)sulfonate, Potassium perfluoro-1-hexanesulfonate, Potassium perfluorohexanesulfonate, Potassium tridecafluorohexan-1-sulfonate, Potassium tridecafluorohexane-1-sulfonate, Tridecafluorohexane-1-sulfonic acid potassium salt, Tridecafluorohexanesulfonic acid potassium salt |
| InChI formel | InChI=1S/C6HF13O3S.K/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22;/h(H,20,21,22);/q;+1/p-1 |
| LEDER | [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | potassium;1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonate |
| Molekylvikt (g/mol) | 438.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?