Läs mer
Tridecafluorohexylethyl Methacrylate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tridecafluorohexylethyl Methacrylate |
| CAS | 2144-53-8 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C12 H9 F13 O2 |
| Kvantitet | 5 g |
| Synonym | 1H,1H,2H,2H-Perfluorooctyl Methacrylate, 2-Propenoic acid','2-methyl-','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (9CI','ACI), 1-Octanol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-','methacrylate (8CI), Methacrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (7CI','8CI), (Perfluorohexyl)ethyl methacrylate, 1H,1H,2H,2H-Perfluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluoro-1-octyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 2-(Perfluorohexyl)ethyl methacrylate, 2-(Tridecafluorohexyl)ethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-yl 2-methyl-2-propenoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl 2-methylprop-2-enoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl methacrylate, 6:2 FTMAC, Actyflon G 06B, C 6FMA, C 6SFMA, Capstone 62MA, Cheminox FAMAC 6, FAMAC 6, Fluowet MA 600, M 1620, MA 600, MAC 6F13, Tridecafluorohexylethyl methacrylate, 6:2 FTMAC, 1H,1H,2H,2H-Perfluorooctyl Methacrylate, 2-Propenoic acid','2-methyl-','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (9CI','ACI), 1-Octanol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-','methacrylate (8CI), Methacrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (7CI','8CI), (Perfluorohexyl)ethyl methacrylate, 1H,1H,2H,2H-Perfluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluoro-1-octyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 2-(Perfluorohexyl)ethyl methacrylate, 2-(Tridecafluorohexyl)ethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-yl 2-methyl-2-propenoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl 2-methylprop-2-enoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl methacrylate, 6:2 FTMAC, Actyflon G 06B, C 6FMA, C 6SFMA, Capstone 62MA, Cheminox FAMAC 6, FAMAC 6, Fluowet MA 600, M 1620, MA 600, MAC 6F13 |
| InChI formel | InChI=1S/C12H9F13O2/c1-5(2)6(26)27-4-3-7(13,14)8(15,16)9(17,18)10(19,20)11(21,22)12(23,24)25/h1,3-4H2,2H3 |
| LEDER | CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate |
| Molekylvikt (g/mol) | 432.18 |
| Visa mer |