missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 97 to 98%, MP Biomedicals™

Catalog Number p-4404900
Click to view available options
Quantity:
100 mL
0.5 L
1 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
9005-65-6
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
604.82
MFCD00082107
HDTIFOGXOGLRCB-KTKRTIGZNA-N
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
9005-65-6
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
604.82
MFCD00082107
HDTIFOGXOGLRCB-KTKRTIGZNA-N
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Triethanolamine ≥97%, Liquid

  • It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks.
  • It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating.
  • It is used in hair care products like hair dyes and wave sets.
  • Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions.
Color Colorless to Yellow
Density 1.124g/cm3 at 20°C
Assay Percent Range 97 to 98%
Quantity 100 mL
Synonym Daltogen, Nitrilo-2, 2, 2-Triethanol
Formula Weight 149.2
Physical Form Liquid
Chemical Name or Material Triethanolamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'