missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 98+%, Thermo Scientific Chemicals

Catalog Number 11371597
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO

Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

Solubility
Miscible with water, methanol, acetone, ether, carbon tetrachloride, n-heptane and chloroform. Slightly soluble in petroleum ether.

Notes
Air sensitive and hygroscopic Incompatible with acids, oxidizing agents, reducing agents and metals.
TRUSTED_SUSTAINABILITY
Melting Point 19°C to 21°C
Density 1.124
Boiling Point 206°C to 207°C (15 mmHg)
Flash Point 185°C (365°F)
Odor Ammonia-like
Linear Formula (HOCH2CH2)3N
Refractive Index 1.485
Quantity 100 g
Beilstein 1699263
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9665
Solubility Information Miscible with water,methanol,acetone,ether,carbon tetrachloride,n-heptane and chloroform. Slightly soluble in petroleum ether.
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material Triethanolamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'