Learn More
Triethanolamine, 99+%, Thermo Scientific Chemicals
Catalog Number 10249510
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1245g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Metal drum
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Refractive Index 1.4830 to 1.4860 (20°C, 589nm)
Quantity 1 L
Fieser 01,1196
Merck Index 15, 9830
Formula Weight 149.19
Percent Purity ≥99.0%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine